Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: GPKDPVKTLR PTPARPRNSL KKATDNEPSQ FSEPRKTLSP TGTAQPSWGV DPKEGPQELQ EKKIQVLEEK VLRLTRTVLD LQSSLAGVSE NLKHATQDDA SRTRAPGLSS
EMILIN2 is a protein coding gene. May be responsible for anchoring smooth muscle cells to elastic fibers, and may be involved not only in the formation of the elastic fiber, but also in the processes that regulate vessel assembly. Has cell adhesive capacity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Elastin microfibril interface-located protein 2; Elastin microfibril interfacer 2; EMILIN-2; extracellular glycoprotein EMILIN-2; FLJ33200; Protein FOAP-10
Gene Aliases: EMILIN-2; EMILIN2; FOAP-10
UniProt ID: (Human) Q9BXX0
Entrez Gene ID: (Human) 84034
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support