Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Control: SiHa cell lysate, 293T cell lysate, human breast carcinoma tissue, mouse testis tissue, SiHa.
Subcellular Location: Cytoskeleton, cytoplasm, lamellipodium, filopodium, synapse, focal adhesion.
This gene encodes a member of the enabled/ vasodilator-stimulated phosphoprotein. Members of this gene family are involved in actin-based motility. This protein is involved in regulating the assembly of actin filaments and modulates cell adhesion and motility. Alternate splice variants of this gene have been correlated with tumor invasiveness in certain tissues and these variants may serve as prognostic markers. A pseudogene of this gene is found on chromosome 3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mammalian enabled variant 11a; mammalian enabled variant pan; NDPP-1; NPC derived proline rich protein 1; NPC-derived proline-rich protein 1; OTTHUMP00000035864; Protein enabled homolog
Gene Aliases: ENA; ENAH; MENA; NDPP-1; NDPP1; WBP8
UniProt ID: (Human) Q8N8S7, (Mouse) Q03173
Entrez Gene ID: (Human) 55740, (Mouse) 13800
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support