Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SINIYLFHKS SYADSVLTHL NLLRQQRLFT DVLLHAGNRT FPCHRAVLAA CSRYFEAMFS GGLKESQDSE VNFDNSIHPE VL
The ectoderm-neural cortex-1 (ENC-1) protein is an early and highly specific marker of neural induction in vertebrates. It is a kelch family related protein that functions as an actin-binding protein and has been suggested to be involved in the organization of the actin cytoskeleton during neural fate specification and development of the nervous system. ENC-1 has also been shown to be required for adipocyte differentiation when cytoskeletal reorganization and cell shape change from fibroblastic preadipocytes to spherical adipocytes occur.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ectoderm-neural cortex protein 1; ectodermal-neural cortex 1 (with BTB domain); ectodermal-neural cortex 1 (with BTB-like domain); ENC-1; FLJ39259; kelch-like 35; kelch-like 37; kelch-like family member 37; Kelch-like protein 37; Nuclear matrix protein NRP/B; nuclear restricted protein, BTB domain-like (brain); p53-induced gene 10 protein; tumor protein p53 inducible protein 10
Gene Aliases: CCL28; ENC-1; ENC1; KLHL35; KLHL37; NRPB; PIG10; TP53I10
UniProt ID: (Human) O14682
Entrez Gene ID: (Human) 8507
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support