Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDVGEEPLEK AARARTAKDP NTYKVLSLVL SVCVLTTILG CIFGLKPSCA KEVKSCKGRC FERTFGNCRC DAACVELGNC CLDYQETCIE PEHIWTCNKF RCGEKRLTRS LCACSDDCKD KGDCCINYSS VCQGEKSWVE EPCESINEPQ CPAGFETPPT LLFSLDGFRA EYLHTWGGLL PVISKLKKCG TYTKNMRPVY PTKTFPNHYS IVTGLYPESH GIIDNKMYDP KMNASFSLKS KEKFNPEWYK GEPIWVTAKY QGLKSGTFFW PGSDVEINGI FPDIYKMYNG SVPFEERILA VLQWLQLPKD ERPHFYTLYL EEPDSSGHSY GPVSSEVIKA LQRVDGMVGM LMDGLKELNL HRCLNLILIS DHGMEQGSCK KYIYLNKYLG DVKNIKVIYG PAARLRPSDV PDKYYSFNYE GIARNLSCRE PNQHFKPYLK HFLPKRLHFA KSDRIEPLTF YLDPQWQLAL NPSERKYCGS GFHGSDNVFS NMQALFVGYG PGFKHGIEAD TFENIEVYNL MCDLLNLTPA PNNGTHGSLN HLLKNPVYTP KHPKEVHPLV QCPFTRNPRD NLGCSCNPSI LPIEDFQTQF NLTVAEEKII KHETLPYGRP RVLQKENTIC LLSQHQFMSG YSQDILMPLW TSYTVDRNDS FSTEDFSNCL YQDFRIPLSP VHKCSFYKNN TKVSYGFLSP PQLNKNSSGI YSEALLTTNI VPMYQSFQVI WRYFHDTLLR KYAEERNGVN VVSGPVFDFD YDGRCDSLEN LRQKRRVIRN QEILIPTHFF IVLTSCKDTS QTPLHCENLD TLAFILPHRT DNSESCVHGK HDSSWVEELL MLHRARITDV EHITGLSFYQ QRKEPVSDIL KLKTHLPTFS QED
This gene is a member of the ecto-nucleotide pyrophosphatase/phosphodiesterase (ENPP) family. The encoded protein is a type II transmembrane glycoprotein comprising two identical disulfide-bonded subunits. This protein has broad specificity and cleaves a variety of substrates, including phosphodiester bonds of nucleotides and nucleotide sugars and pyrophosphate bonds of nucleotides and nucleotide sugars. This protein may function to hydrolyze nucleoside 5' triphosphates to their corresponding monophosphates and may also hydrolyze diadenosine polyphosphates. Mutations in this gene have been associated with 'idiopathic' infantile arterial calcification, ossification of the posterior longitudinal ligament of the spine (OPLL), and insulin resistance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alkaline phosphodiesterase 1; E-NPP 1; Ectonucleotide pyrophosphatase/phosphodiesterase family member 1; HGNC:3356; Ly-41 antigen; Membrane component chromosome 6 surface marker 1; membrane component, chromosome 6, surface marker 1; OTTHUMP00000043194; Phosphodiesterase I/nucleotide pyrophosphatase 1; plasma-cell membrane glycoprotein 1; Plasma-cell membrane glycoprotein PC-1
Gene Aliases: ARHR2; COLED; ENPP1; M6S1; NPP1; NPPS; PC-1; PC1; PCA1; PDNP1
UniProt ID: (Human) P22413
Entrez Gene ID: (Human) 5167
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support