Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RKLLVFLLDG FRSDYISDEA LESLPGFKEI VSRGVKVDYL TPDFPSLSYP NYYTLMTGRH CEVHQMIGNY MWDPTTNKSF DIGVNKDSLM PLWWNGS
Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
Choline-specific glycerophosphodiester phosphodiesterase. Hydrolyzes the classical substrate for phospholipase C, p-nitrophenyl phosphorylcholine, while it does not hydrolyze the classical nucleotide phosphodiesterase substrate, p-nitrophenyl thymidine 5'-monophosphate. Hydrolyzes lysophosphatidylcholine (LPC) to form monoacylglycerol and phosphorylcholine but not lysophosphatidic acid, showing it has a lysophospholipase C activity. Has a preference for LPC with short (12:0 and 14:0) or polyunsaturated (18:2 and 20:4) fatty acids. Also hydrolyzes glycerophosphorylcholine and sphingosylphosphorylcholine efficiently.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B830047L21Rik; Choline-specific glycerophosphodiester phosphodiesterase; E-NPP 6; Ectonucleotide pyrophosphatase/phosphodiesterase family member 6; glycerophosphocholine cholinephosphodiesterase; Glycerophosphocholine cholinephosphodiesterase ENPP6; GPC-Cpde; NPP-6
Gene Aliases: ENPP6; NPP6; UNQ1889/PRO4334
UniProt ID: (Human) Q6UWR7
Entrez Gene ID: (Human) 133121
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support