Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FNELDLETL APYIPMDGED FQLSPICPEE RLLAENPQST PQHCFSAMTN IFQPLAPVAP HSPFLLDKFQ QQLESKKTEP EHRPMSSIFF DAGSKASLPP CCGQASTPLS SMGGRSNTQW PPDPPLHFGP TKWAVGDQRT EFLGAAPLGP PVSPPHVSTF KTRSAKGFGA RGPDVLSPAM VALSNKLKLK RQLEYEEQAF QDLSGGDPPG GSTSHLMWKR MKNLRGGSCP LMPDKPLSAN VPNDKFTQNP MRGLGHPLRH LPLPQPPSAI SPGENSKSRF PPQCYATQYQ DYSLSSAHKV SGMASRLLGP SFESYLLPEL TRYDCEVNVP VLGSSTLLQG GDLLRALDQA T (521-870 aa encoded by BC051338)
HIF-2 alpha (Epas1) is a transcription factor involved in the induction of genes regulated by oxygen. HIF-2 alpha contains a basic-helix-loop-helix domain protein dimerization domain as well as a domain found in proteins in signal transduction pathways which respond to oxygen levels. It also regulates the vascular endothelial growth factor (VEGF) expression and seems to be implicated in the development of blood vessels and the tubular system of the lungs. Mutations in this gene are associated with erythrocytosis familial type 4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basic-helix-loop-helix-PAS protein MOP2; bHLHe73; Class E basic helix-loop-helix protein 73; Endothelial PAS domain-containing protein 1; EPAS-1; HIF-1-alpha-like factor; HIF-1alpha-like factor; HIF-2-alpha; HIF2-alpha; HLF; hypoxia-inducible factor 2 alpha; Hypoxia-inducible factor 2-alpha; Member of PAS protein 2; PAS domain-containing protein 2
Gene Aliases: BHLHE73; ECYT4; EPAS1; HIF2A; HLF; MOP2; PASD2
UniProt ID: (Human) Q99814
Entrez Gene ID: (Human) 2034
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support