Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DACQACSPGF FKFEASESPC LECPEHTLPS PEGATSCECE EGFFRAPQDP ASMPCTRPPS APHYLTAVGM GAKVELRWTP PQDSGGREDI VYSVTCEQCW PESGECGPCE ASVRYSEPPH GLTRTSVTVS DLEPHMNYTF TVEARNGVSG LVTSRSFRTA SVSINQTEPP KVRLEGRSTT SLSVSWSIPP PQQSRVWKYE VTYRKKGDSN SYNVRRTEGF SVTLDDLAPD TTYLVQVQAL TQEGQGAGSK VHEFQTLSPE GSGNL
EPHA2 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA2 is overexpressed in many types of human cancers. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ephrin type-A receptor 2; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; soluble EPHA2 variant 1; Tyrosine-protein kinase receptor ECK
Gene Aliases: ARCC2; CTPA; CTPP1; CTRCT6; ECK; EPHA2
UniProt ID: (Human) P29317
Entrez Gene ID: (Human) 1969
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support