Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FSTLTLLLGQ RFGRFLGLTE RDVELLYPVK EKVFYSLMRE SGYMHIQCTK PDTVGSALND SPVGLAAYIL EKFSTWTNTE FRYLEDGGLE RKFSLDDLLT NVMLYWTTGT IISSQRFYKE NLGQGWMTQK HERMKVYVPT GFSAFPFELL HTPEKWVRFK YPKLISYSYM VRGGHFAAFE EPELLAQDIR KFLSVLERQ
Biotransformation enzyme that catalyzes the hydrolysis of arene and aliphatic epoxides to less reactive and more water soluble dihydrodiols by the trans addition of water. May play a role in the metabolism of endogenous lipids such as epoxide-containing fatty acids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epoxide hydratase; Epoxide hydrolase 1; epoxide hydrolase 1 microsomal; epoxide hydrolase 1, microsomal (xenobiotic); mEH; Microsomal epoxide hydrolase
Gene Aliases: AI195553; Eph-1; Eph1; EPHX; EPHX1; EPOX; Epxh1; HYL1; MEH
UniProt ID: (Human) P07099, (Mouse) Q9D379
Entrez Gene ID: (Human) 2052, (Mouse) 13849
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support