Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Acts as GTPase-activating protein for RAB27A, but not for RAB2A, RAB3A, nor RAB4A. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AC004997.C22.2; EBP50-PDX interactor of 64 kDa; EBP50-PDZ interactor of 64 kD; EPI64 protein; Rab27A-GAP-alpha; TBC1 domain family member 10A; TBC1 domain family, member 10A
Gene Aliases: AI447804; dJ130H16.1; dJ130H16.2; EPI64; mFLJ00288; TBC1D10; TBC1D10A
UniProt ID: (Human) Q9BXI6, (Mouse) P58802
Entrez Gene ID: (Human) 83874, (Mouse) 103724
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support