Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VHECPAWLW LLLSLLSLPL GLPVLGAPPR LICDSRVLER YLLEAKEAEN ITTGCAEHCS LNENITVPDT KVNFYAWKRM EVGQQAVEVW QGLALLSEAV LRGQALLVNS SQPWEPLQLH VDKAVSGLRS LTTLLRALGA QKEAISPPDA ASAAPLRTIT ADTFRKLFRV YSNFLRGKLK LYTGEACRTG DR (3-193 aa encoded by BC093628)
Erythropoietin (EPO) protein is a growth factor essential for the survival, proliferation, and differentiation of erythroid precursors. EPO is not responsible for early commitment to the lineage, and instead induces the final maturation of partially differentiated erythroid cells and the production of red cell-specific proteins, such as hemoglobin. The EPO receptor (EPOR) is present in higher levels on precursor cells, and expression diminishes with cell maturation.Hepatocytes are the main source of EPO in the fetus, followed by interstitial cells of the kidney after birth and through adulthood. EPO expression is upregulated during hypoxia, and low levels of transcription have also been detected in lung and brain tissue under these conditions. Recent data suggest that EPO may also play a role in the protection of neuronal tissue from damage and initiation of repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epoetin; Erythropoietin; MGC138142
Gene Aliases: EP; EPO; MVCD2
UniProt ID: (Human) P01588
Entrez Gene ID: (Human) 2056
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support