Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: Raji, K562, BCL-1
Predicted Reactivity: Chicken (100%), Bovine (100%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EPS15 is a ubiquitously expressed protein that is involved in cell growth regulation and may be involved in the regulation of mitogenic signals and control of cell proliferation. Phosphorylation of EPS15 on Tyrosine 849 upon DNA damage is involved in the internalization of EGFR. A chromosomal aberration involving EPS15 is also found in acute leukemias, resulting in a rogue activator protein.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AF-1p protein; ALL1 fused gene from chromosome 1; epidermal growth factor pathway substrate 15; Epidermal growth factor receptor substrate 15; Protein AF-1p; Protein Eps15
Gene Aliases: 2410112D09Rik; AF-1P; AF1P; EPS15; MLLT5
UniProt ID: (Human) P42566, (Mouse) P42567
Entrez Gene ID: (Human) 2060, (Mouse) 13858
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support