Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MHLLPALAGV LATLVLAQPC EGTDPASPGA VETSVLRDCI AEAKLLVDAA YNWTQKSIKQ RLRSGSASPM DLLSYFKQPV AATRTVVRAA DYMHVALGLL EEKLQPQRSG PFNVTDVLTE PQLRLLSQAS GCALRDQAER CSDKYRTITG RCNNKRRPLL GASNQALARW LPAEYEDGLS LPFGWTPSRR RNGFLLPLVR AVSNQIVRFP NERLTSDRGR ALMFMQWGQF IDHDLDFSPE SPARVAFTAG VDCERTCAQL PPCFPIKIPP NDPRIKNQRD CIPFFRSAPS CPQNKNRVRN QINALTSFVD ASMVYGSEVS LSLRLRNRTN YLGLLAINQR FQDNGRALLP FDNLHDDPCL LTNRSARIPC FLAGDTRSTE TPKLAAMHTL FMREHNRLAT ELRRLNPRWN GDKLYNEARK IMGAMVQIIT YRDFLPLVLG KARARRTLGH YRGYCSNVDP RVANVFTLAF RFGHTMLQPF MFRLDSQYRA SAPNSHVPLS SAFFASWRIV YEGGIDPILR GLMATPAKLN RQDAMLVDEL RDRLFRQVRR IGLDLAALNM QRSRDHGLPG YNAWRRFCGL SQPRNLAQLS RVLKNQDLAR KFLNLYGTPD NIDIWIGAIA EPLLPGARVG PLLACLFENQ FRRARDGDRF WWQKRGVFTK RQRKALSRIS LSRIICDNTG ITTVSRDIFR ANIYPRGFVN CSRIPRLNLS AWRGT
EPX belongs to the peroxidase family, XPO subfamily. Defects in EPX are the cause of eosinophil peroxidase deficiency (EPD).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Eosinophil peroxidase; EPO; EPX PEN; PERE
Gene Aliases: EPER; EPO; EPP; EPX; EPX-PEN; EPXD
UniProt ID: (Human) P11678
Entrez Gene ID: (Human) 8288
Molecular Function:
peroxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support