Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LPQLSDDIPF RVNWPGTEFS LPTTGVLYKE DNYVIMTTAH KEKYKCILPL VTSGDEEEEK DYKGPNPREL LEPLFKQSSC SYRIESYWTY EVCHGKHIRQ YHEEKETGQK INIHEYYLGN MLAKNLLFEK EREAEEKEKS NEIPTKNIEG QM
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer invasion and metastasis-related; Endoplasmic reticulum lectin 1; epididymis luminal protein 117; ER lectin; Erlectin; erlectin 1; XTP3-transactivated gene B protein; XTP3-transactivated protein B
Gene Aliases: 4933407N01Rik; C2orf30; CIM; CL24936; CL25084; ERLEC1; HEL117; RGD1306508; UNQ1878/PRO4321; XTP3-B; XTP3TPB
UniProt ID: (Human) Q96DZ1, (Mouse) Q8VEH8
Entrez Gene ID: (Human) 27248, (Mouse) 66753, (Rat) 289874
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support