Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSIAGVAAQE IRVPLKTGFL HNGRAMGNMR KTYWSSRSEF KNNFLNIDPI TMAYSLNSSA QERLIPLGHA SKSAPMNGHC FAENGPSQKS SLPPLLIPPS ENLGPHEEDQ VVCGFKKLTV NGVCASTPPL TPIKNSPSLF PCAPLCERGS RPLPPLPISE ALSLDDTDCE VEFLTSSDTD FLLEDSTLSD FKYDVPGRRS FRGCGQINYA YFDTPAVSAA DLSYVSDQNG GVPDPNPPPP QTHRRLRRSH SGPAGSFNKP AIRISNCCIH RASPNSDEDK PEVPPRVPIP PRPVKPDYRR WSAEVTSSTY SDEDRPPKVP PREPLSPSNS RTPSPKSLPS YLNGVMPPTQ SFAPDPKYVS SKALQRQNSE GSASKVPCIL PIIENGKKVS STHYYLLPER PPYLDKYEKF FREAEETNGG AQIQPLPADC GISSATEKPD SKTKMDLGGH VKRKHLSYVV SP
ERRFI1 is a cytoplasmic protein whose expression is upregulated with cell growth (Wick et al., 1995). It shares significant homology with the protein product of rat gene-33, which is induced during cell stress and mediates cell signaling (Makkinje et al., 2000).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERBB receptor feedback inhibitor 1; MIG-6; Mitogen-inducible gene 6 protein; receptor-associated late transducer
Gene Aliases: ERRFI1; GENE-33; MIG-6; MIG6; RALT
UniProt ID: (Human) Q9UJM3
Entrez Gene ID: (Human) 54206
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support