Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAALTPRKR KQDSLKCDSL LHFTENLFPS PNKKHCFYQN SDKNEENLHC SQQEHFVLSA LKTTEINRLP SANQGSPFKS ALSTVSFYNQ NKWYLNPLER KLIKESRSTC LKTNDEDKSF PIVTEKMQGK PVCSKKNNKK PQKSLTAKYQ PKYRHIKPVS RNSRNSKQNR VIYKPIVEKE NNCHSAENNS NAPRVLSQKI KPQVTLQGGA AFFVRKKSSL RKSSLENEPS LGRTQKSKSE VIEDSDVETV SEKKTFATRQ VPKCLVLEEK LKIGLLSASS KNKEKLIKDS SDDRVSSKEH KVDKNEAFSS EDSLGENKTI SPKSTVYPIF SASSVNSKRS LGEEQFSVGS VNFM (1-353 aa encoded by BC146562)
This gene encodes a protein that may have acetyltransferase activity and may be required for the establishment of sister chromatid cohesion during the S phase of mitosis. Mutations in this gene have been associated with Roberts syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ECO1 homolog 2; EFO2; Establishment factor-like protein 2; Establishment of cohesion 1 homolog 2; establishment of sister chromatid cohesion N-acetyltransferase 2; N-acetyltransferase ESCO2
Gene Aliases: 2410004I17Rik; D030072L07Rik; EFO2; ESCO2; RBS
UniProt ID: (Human) Q56NI9, (Mouse) Q8CIB9
Entrez Gene ID: (Human) 157570, (Mouse) 71988, (Rat) 691979
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support