Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MFRAAAPGQ LRRAASLLRF QSTLVIAEHA NDSLAPITLN TITAATRLGG EVSCLVAGTK CDKVAQDLCK VAGIAKVLVA QHDVYKGLLP EELTPLILAT QKQFNYTHIC AGASAFGKNL LPRVAAKLEV APISDIIAIK SPDTFVRTIY AGNALCTVKC DEKVKVFSVR GTSFDAAATS GGSASSEKAS STSPVEISEW LDQKLTKSDR PELTGAKVVV SGGRGLKSGE NFKLLYDLAD QLHAAVGASR AAVDAGFVPN DMQVGQTGKI VAPELYIAVG ISGAIQHLAG MKDSKTIVAI NKDPEAPIFQ VADYGIVADL FKVVPEMTEI LKKK (1-333 aa encoded by BC015526)
ETFA participates in catalyzing the initial step of the mitochondrial fatty acid beta-oxidation. It shuttles electrons between primary flavoprotein dehydrogenases and the membrane-bound electron transfer flavoprotein ubiquinone oxidoreductase. Defects in electron-transfer-flavoprotein have been implicated in type II glutaricaciduria in which multiple acyl-CoA dehydrogenase deficiencies result in large excretion of glutaric, lactic, ethylmalonic, butyric, isobutyric, 2-methyl-butyric, and isovaleric acids. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-ETF; electron transfer flavoprotein alpha-subunit; Electron transfer flavoprotein subunit alpha, mitochondrial; electron transfer flavoprotein, alpha polypeptide; electron transferring flavoprotein, alpha polypeptide; electron-transfer-flavoprotein alpha polypeptide; glutaric aciduria II
Gene Aliases: 2010200I21Rik; D9Ertd394e; EMA; ETF; ETFA; GA2; MADD
UniProt ID: (Human) P13804, (Mouse) Q99LC5, (Rat) P13803
Entrez Gene ID: (Human) 2108, (Mouse) 110842, (Rat) 300726
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support