Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LVALGLVVGL DYQNPYLSPF REFQRWKHHP SIRPTLEGGK RIAYGARALN EGGFQSIPKL TFPGGLLIGC SPGFMNVPKI KGTHTAMKSG ILAAESIFNQ LTSENLQSKT IGLHVTEYED NLKNSWVWKE LYSVRNIRPS CHGVLGVYGG MIYTGIFYWI LRGMEPWTLK HKGSDFERLK PAKDCTPIEY PKPDGQISFD LLSSVALSGT NHEHDQPAHL TLRDDSIPVN RNLSIYDGPE QRFCPAGVYE FVPVEQGDGF RLQINAQNCV HCKTCDIKDP SQNINWVVPE GGGGPAYNGM
Electron-transferring-flavoprotein dehydrogenase in the inner mitochondrial membrane accepts electrons from electron-transfer flavoprotein which is located in the mitochondrial matrix and reduces ubiquinone in the mitochondrial membrane. The protein is synthesized as a 67-kDa precursor which is targeted to mitochondria and processed in a single step to a 64-kDa mature form located in the mitochondrial membrane. Deficiency in electron-transferring-flavoprotein dehydrogenase have been demonstrated in some patients with type II glutaricacidemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Electron transfer flavoprotein-ubiquinone oxidoreductase, mitochondrial; electron transferring flavoprotein dehydrogenase; Electron-transferring-flavoprotein dehydrogenase; ETF dehydrogenase; ETF-QO; ETF-ubiquinone oxidoreductase
Gene Aliases: 0610010I20Rik; AV001013; ETFDH; ETFQO; MADD
UniProt ID: (Human) Q16134, (Mouse) Q921G7
Entrez Gene ID: (Human) 2110, (Rat) 295143, (Mouse) 66841
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support