Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VARRQLSQR GGSGAPILLR QMFEPVSCTF TYLLGDRESR EAVLIDPVLE TAPRDAQLIK ELGLRLLYAV NTHCHADHIT GSGLLRSLLP GCQSVISRLS GAQADLHIED GDSIRFGRFA LETRAS (8-132 aa encoded by B C008250)
ETHE1 is a sulfur dioxygenase that localizes within the mitochondrial matrix. The enzyme functions in sulfide catabolism. Mutations in its gene result in ethylmalonic encephalopathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ethylmalonic encephalopathy 1; Ethylmalonic encephalopathy protein 1; Ethylmalonic encephalopathy protein 1 homolog; Hepatoma subtracted clone one protein; Persulfide dioxygenase ETHE1, mitochondrial; protein ETHE1, mitochondrial; Sulfur dioxygenase ETHE1
Gene Aliases: 0610025L15Rik; ETHE1; HSCO; YF13H12
UniProt ID: (Human) O95571, (Mouse) Q9DCM0
Entrez Gene ID: (Human) 23474, (Mouse) 66071
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support