Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKAAVDLKPT LTIIKTEKVD LELFPSPDME CADVPLLTPS SKEMMSQALK ATFSGFTKEQ QRLGIPKDPR QWTETHVRDW VMWAVNEFSL KGVDFQKFCM NGAALCALGK DCFLELAPDF VGDILWEHLE ILQKEDVKPY QVNGVNPAYP ESRYTSDYFI SYGIEHAQCV PPSEFSEPSF ITESYQTLHP ISSEELLSLK YENDYPSVIL RDPLQTDTLQ NDYFAIKQEV VTPDNMCMGR TSRGKLGGQD SFESIESYDS CDRLTQSWSS QSSFNSLQRV PSYDSFDSED YPAALPNHKP KGTFKDYVRD RADLNKDKPV IPAAALAGYT GSGPIQLWQF LLELLTDKSC QSFISWTGDG WEFKLSDPDE VARRWGKRKN KPKMNYEKLS RGLRYYYDKN IIHKTAGKRY VYRFVCDLQS LLGYTPEELH AMLDVKPDAD E
ETS1 (c-Ets-1; v-ets erythroblastosis virus E26 oncogene homlog 1 (avian); ETS-1; EWSR2; p54), a 441 A. A protein, is a member of DNA-binding ETS protein family. It is a multifunctional protein consisting of an N-terminal pointed domain, a central transactivation domain and 2 auto inhibitory domains flanked on both sides of conserved C-terminal Ets domain. It interacts with factors including PEBP2alphaA, p300/CBP and mediates transactivation of Osteopontin, Presenilin-1, Stromelysin and Collagenase. It is activated by Ras/MAPK signaling and its expression is up-regulated in resting T-cells. Its activity is modulated by various factors including AP-1, Sp-1, c-myb and MafB. Sp100 activates ETS1 in nuclear bodies. It interacts with EAP1/Daxx and causes repression of MMP1 and BCL2 transactivation. It interacts with STAT6 and modulates Socs-1 expression induced by IL-4. It interacts with GATA3 and positively regulates Tax-1 dependent IL-5 expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1; avian erythroblastosis virus E26 oncogene 1; ets protein; Ets-1Delta(III-VI); FLJ10768; p54; Protein C-ets-1; tumor progression locus 1; v-ets avian erythroblastosis virus E2 oncogene homolog 1; v-ets avian erythroblastosis virus E26 oncogene homolog 1
Gene Aliases: AI196000; AI448617; c-ets-1; D230050P06; ETS-1; ETS1; EWSR2; p54; Tpl1
UniProt ID: (Human) P14921, (Mouse) P27577
Entrez Gene ID: (Human) 2113, (Mouse) 23871
Molecular Function: winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support