Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MATSEQSIC QARASVMVYD DTSKKWVPIK PGQQGFSRIN IYHNTASNTF RVVGVKLQDQ QVVINYSIVK GLKYNQATPT FHQWRDARQV YGLNFASKEE ATTFSNAMLF ALNIMNSQEG GPSSQRQVQN GPSPDEMDIQ RRQVMEQHQQ QRQESLERRT SATGPILPPG HPSSAASAPV SCSGPPPPPP PPVPPPPTGA TPPPPPPLPA GGAQGSSHDE SSMSGLAAAI AGAKLRRVQR PEDASGGSSP SGTSKSDANR ASSGGGGGGL MEEMNKLLAK RRKAASQSDK PAEKKEDESQ MEDPSTSPSP GTRAASQPPN SSEAGRKPWE RSNSVEKPVS SILSRTPSVA KSPEAKSPLQ SQPHSRMKPA GSVNDMALDA FDLDRMKQEI LEEVVRELHK VKEEIIDAIR QELSGISTT (1-418 aa encoded by BC032358 )
Ena/VASP proteins are actin-associated proteins involved in a range of processes dependent on cytoskeleton remodeling and cell polarity such as axon guidance and lamellipodial and filopodial dynamics in migrating cells. EVL enhances actin nucleation and polymerization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ena/vasodilator-stimulated phosphoprotein-like; Ena/VASP-like protein
Gene Aliases: EVL; RNB6
UniProt ID: (Human) Q9UI08
Entrez Gene ID: (Human) 51466
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support