Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MKETPLSNCE RRFLLRAIEE KKRLDGRQTY DYRNIRISFG TDYGCCIVEL GKTRVLGQVS CELVSPKLNR ATEGILFFNL ELSQMAAPAF EPGRQSDLLV KLNRLMERCL RNSKCIDTES LCVVAGEKVW QIRVDLHLLN HDGNIIDAAS IAAIVALCHF RRPDVSVQGD EVTLYTPEER DPVPLSIHHM PICVSFAFFQ QGAYLLVDPN EREERVMDGL LVIAMNKHRE ICTIQSSGGI MLLKDQVLRC SKIAGVKVAE ITELILKALE NDQKVRKEGG KFGFAESIAN QRITAFKMEK APIDTSDVEE KAEEIIAEAE PPSEVVSTPV LWTPGTAQIG EGVENSWGDL E
This gene encodes a component of the human exosome, a exoribonuclease complex which processes and degrades RNA in the nucleus and cytoplasm. This component may play a role in mRNA degradation and the polymyositis/scleroderma autoantigen complex. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Autoantigen PM/Scl 1; Exosome complex component RRP45; exosome complex exonuclease RRP45; Exosome component 9; P75 polymyositis-scleroderma overlap syndrome associated autoantigen; P75 polymyositis-scleroderma overlap syndrome-associated autoantigen; PM/Scl-75; PMSCL autoantigen, 75kD; Polymyositis/scleroderma autoantigen 1; polymyositis/scleroderma autoantigen 1, 75kDa; Polymyositis/scleroderma autoantigen 75 kDa
Gene Aliases: EXOSC9; p5; p6; PM/Scl-75; PMSCL1; RRP45; Rrp45p
UniProt ID: (Human) Q06265, (Mouse) Q9JHI7, (Rat) Q4QR75
Entrez Gene ID: (Human) 5393, (Mouse) 50911, (Rat) 294975
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support