Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
EXTL2 is a glycosyltransferase required for the biosynthesis of heparan-sulfate and responsible for the alternating addition of beta-1-4-linked glucuronic acid (GlcA) and alpha-1-4-linked N-acetylglucosamine (GlcNAc) units to nascent heparan sulfate chains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-1,4-N-acetylhexosaminyltransferase EXTL2; Alpha-GalNAcT EXTL2; exostoses (multiple)-like 2; exostoses-like 2; Exostosin-like 2; exostosin-like glycosyltransferase 2; exotoses (multiple)-like 2; EXT-related protein 2; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; processed exostosin-like 2
Gene Aliases: 3000001D04Rik; AW146439; EXTL2; EXTR2
UniProt ID: (Human) Q9UBQ6, (Mouse) Q9ES89
Entrez Gene ID: (Human) 2135, (Rat) 310803, (Mouse) 58193
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support