Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEMQDLTSP HSRLSGSSES PSGPKLGNSH INSNSMTPNG TEVKTEPMSS SETASTTADG SLNNFSGSAI GSSSFSPRPT HQFSPPQIYP SNRPYPHILP TPSSQTMAAY GQTQFTTGMQ QATAYATYPQ PGQPYGISSY GALWAGIKTE GGLSQSQSPG QTGFLSYGTS FSTPQPGQAP YSYQMQGSSF TTSSGIYTGN NSLTNSSGFN SSQQDYPSYP SFGQGQYAQY YNSSPYPAHY MTSSNTSPTT PSTNATYQLQ EPPSGITSQA VTDPTAEYST IHSPSTPIKD SDSDRLRRGS D (1-300 aa encoded by BC121799)
This gene encodes a member of the eyes absent family of proteins. The encoded protein may play a role in the developing kidney, branchial arches, eye, and ear. Mutations of this gene have been associated with branchiootorenal dysplasia syndrome, branchiootic syndrome, and sporadic cases of congenital cataracts and ocular anterior segment anomalies. A similar protein in mice can act as a transcriptional activator. Four transcript variants encoding three distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Eyes absent; Eyes absent homolog 1; MGC141875; OTTHUMP00000195053
Gene Aliases: BOP; BOR; BOS1; EYA1; OFC1
UniProt ID: (Human) Q99502, (Mouse) P97767
Entrez Gene ID: (Human) 2138, (Mouse) 14048
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support