Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: HVVVIQNQTL IKAYINNSLI LSEDIDPHKN FVALNYDGIC YLGGFEYGRK VNIVTQEIFK TNFVGKIKDV VFFQEPKNIE LIKLEGYNVY DGDEQNEVT
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EGF-like protein 10; EGF-like protein 11; EGF-like-domain, multiple 10; EGF-like-domain, multiple 11; Epidermal growth factor-like protein 10; Epidermal growth factor-like protein 11; Protein eyes shut homolog; Protein spacemaker homolog
Gene Aliases: bA166P24.2; bA307F22.3; bA74E24.1; C6orf178; C6orf179; C6orf180; dJ1018A4.2; dJ22I17.2; dJ303F19.1; EGFL10; EGFL11; EYS; RP25; SPAM; UNQ9424/PRO34591
UniProt ID: (Human) Q5T1H1
Entrez Gene ID: (Human) 346007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support