Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MPKPINVRVT TMDAELEFAI QPNTTGKQLF DQVVKTIGLR EVWYFGLHYV DNKGFPTWLK LDKKVSAQEV RKENPLQFKF RAKFYPEDVA EELIQDITQK LFFLQVKEGI LSDEIYCPPE TAVLLGSYAV QAKFGDYNKE VHKSGYLSSE RLIPQRVMDQ HKLTRDQWED RIQVWHAEHR GMLKDNAMLE YLKIAQDLEM YGINYFEIKN KKGTDLWLGV DALGLNIYEK DDKLTPKIGF PWSEIRNISF NDKKFVIKPI DKKAPDFVFY APRLRINKRI LQLCMGNHEL YMRRRKPDTI EVQQMKAQAR EEKHQKQLER QQLETEKKRR ETVEREKEQM MREKEELMLR LQDYEEKTKK AERELSEQIQ RALQLEEERK RAQEEAERLE ADRMAALRAK EELERQAVDQ IKSQEQLAAE LAEYTAKIAL LEEARRRKED EVEEWQHRAK EAQDDLVKTK EELHLVMTAP PPPPPPVYEP VSYHVQESLQ DEGAEPTGYS AELSSEGIRD DRNEEKRITE AEKNERVQRQ LLTLSSELSQ ARDENKRTHN DIIHNENMRQ GRDKYKTLRQ IRQGNTKQRI DEFEAL
The cytoplasmic peripheral membrane protein encoded by this gene functions as a protein-tyrosine kinase substrate in microvilli. As a member of the ERM protein family, this protein serves as an intermediate between the plasma membrane and the actin cytoskeleton. This protein plays a key role in cell surface structure adhesion, migration and organization, and it has been implicated in various human cancers. A pseudogene located on chromosome 3 has been identified for this gene. Alternatively spliced variants have also been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytovillin; cytovillin 2; DKFZp762H157; epididymis secretory protein Li 105; EZRI; Ezrin; FLJ26216; MGC1584; p81; villin 2 (ezrin); Villin-2
Gene Aliases: CVIL; CVL; EZR; HEL-S-105; VIL2
UniProt ID: (Human) P15311
Entrez Gene ID: (Human) 7430
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support