Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Endotoxin level < 0.01EU/µg by LAL method
Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Emapalumab, also known as NI-0501, is a fully human monoclonal antibody that targets interferon gamma. Emapalumab development was sponsored by NovImmune SA, further developed by Sobi and FDA approved on November 20, 2018. 1, 5 The approval of emapalumab was followed by the designation of orphan pharmaceutical, priority review and breakthrough therapy. 5 As well, emapalumab was given the status of PRIME by the EMA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: emapalumab-lzsg; NI-0501
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support