Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Endomucin (EMCN) also known as endothelial sialomucin or mucin-like sialoglycoprotein interferes with the assembly of focal adhesion complexes and inhibits interaction between cells and the extracellular matrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endomucin; endomucin-1; Endomucin-1/2; Endomucin-2; Gastric cancer antigen Ga34; MUC-14; mucin 14, endothelial; Mucin-14
Gene Aliases: 0610012K22Rik; AI315669; EMCN; EMCN2; MUC14
UniProt ID: (Human) Q9ULC0, (Mouse) Q9R0H2
Entrez Gene ID: (Human) 51705, (Mouse) 59308
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support