Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-20534 detects Endothelin B Receptor in human samples.
Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AU022549; Endothelin receptor non-selective type; endothelin receptor subtype B1; Endothelin receptor type B; ET-B; Et-b receptor; Et-b-r; ET-RB; Etb receptor; Etb-svr; RP11-318G21.1
Gene Aliases: ABCDS; EDNRB; ET-B; ET-BR; ETB; ETB1; ETBR; ETRB; HSCR; HSCR2; WS4A
UniProt ID: (Human) Q8NHM6
Entrez Gene ID: (Human) 1910
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support