Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 77%
ENDOV gene ontology annotations related to this gene include DNA repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endonuclease V; novel endonuclease V protein; putative endonuclease FLJ39025 homolog
Gene Aliases: A730011L01Rik; Endov
UniProt ID: (Mouse) Q8C9A2
Entrez Gene ID: (Mouse) 338371
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support