Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 12
Enkurin interacts with transient receptor potential canonical.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Enkurin
Gene Aliases: C10orf63; CFAP106; ENKUR
UniProt ID: (Human) Q8TC29
Entrez Gene ID: (Human) 219670
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support