Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: enactin; Entactin; entactin 1; NID 1; NID-1; Nidogen (enactin); nidogen (entactin); Nidogen precursor (Entactin); Nidogen-1; Nidogen-1;
Gene Aliases: A630025O17; Ent; entactin; entactin-1; NID; NID1; nidogen-1
UniProt ID: (Human) P14543, (Mouse) P10493
Entrez Gene ID: (Pig) 100511351, (Human) 4811, (Rat) 25494, (Mouse) 18073
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support