Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
AA Sequence of recombinant protein: VTIPSSCCTS FISKKIPENR VVSYQLANGS ICPKAGVIFI TKKGHKICTD PKLLWVQRHI QKLDAKKNQP SKGAKAVRTK FAVQRRRGNS TEV.
Preparation: Produced from sera of goats immunized with highly pure Recombinant Murine Eotaxin-2 (CCL24). Anti-Murine Eotaxin-2 (CCL24)-specific antibody was purified by affinity chromatography and then biotinylated.
Sandwich ELISA: To detect mEotaxin-2 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine Eotaxin-2 (500-P175G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mEotaxin-2.
Western Blot: To detect mEotaxin-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mEotaxin-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
500-P175GBT-1MG will be provided as 2 x 500 µg
Eotaxin 2 belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes, a minimal activity on neutrophils, and is negative on monocytes and activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C motif chemokine 24; CC chemokine CCL24; Eosinophil chemotactic protein 2; Eotaxin-2; Eotaxin2; small inducible cytokine A24; Small-inducible cytokine A24
Gene Aliases: Ccl24; CKb-6; MPIF-2; Scya24
UniProt ID: (Mouse) Q9JKC0
Entrez Gene ID: (Mouse) 56221
Molecular Function: cytokine intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support