Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This affinity purified antibody is directed against human Ephrin B1, B2, B3. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with Ephrin-B1 to human, mouse, rat, and Gallus gallus; 100% homology with Ephrin-B2 to mouse; 100% homology with Ephrin-B3 to human and mouse.
Eph receptors and ephrins exhibit complementary expression in many tissues during embryogenesis indicating that bidirectional activation of Eph receptors and ephrin-B proteins may occur at expression domain interfaces. Ephrin-B1 transduces outside-in signals through C-terminal protein interactions that effect integrin-mediated cell attachment and migration. The distribution of ephrin-B1 in the developing retina suggests that it influences retinal axon mapping along the dorsal-ventral axis and may be involved in intratectal development The transmembrane ligand ephrin-B2 and its receptor tyrosine kinase EphB4 are specifically expressed on arterial and venous endothelial cells, respectively.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CEK5 receptor ligand; CEK5-L; EFL-3; ELF-2; ELK ligand; ELK-L; EPH-related receptor transmembrane ligand ELK-L3; EPH-related receptor tyrosine kinase ligand 2; EPH-related receptor tyrosine kinase ligand 5; EPH-related receptor tyrosine kinase ligand 8; Ephrin-B1; Ephrin-B2; Ephrin-B3; HTK ligand; HTK-L; LERK-2; LERK-5; LERK-8; ligand of eph-related kinase 5; Stimulated by retinoic acid gene 1 protein
Gene Aliases: Cek5-L; CFND; CFNS; EFB1; EFL-3; EFL-6; EFL3; EFL6; EFNB1; EFNB2; EFNB3; ELF-2; ELF-3; Elf2; Elk-L; Elk-L3; Epl2; Epl5; Epl8; EPLG2; EPLG5; EPLG8; Htk-L; HTKL; LERK-2; LERK-5; LERK-8; LERK2; LERK5; LERK8; NLERK-1; NLERK-2; Stra1
UniProt ID: (Human) P98172, (Human) P52799, (Human) Q15768, (Mouse) P52795, (Mouse) P52800, (Mouse) O35393
Entrez Gene ID: (Human) 1947, (Human) 1948, (Human) 1949, (Mouse) 13641, (Mouse) 13642, (Mouse) 13643
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support