Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Immunogen sequence: MAVRRDSVWK YCWGVLMVLC RTAISKSIVL EPIYWNSSNS KFLPGQGLVL YPQIGDKLDI ICPKVDSKTV GQYEYYKVYM VDKDQADRCT IKKENTPLLN
This gene encodes a member of the ephrin (EPH) family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. This gene encodes an EFNB class ephrin which binds to the EPHB4 and EPHA3 receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ELF-2; EPH-related receptor tyrosine kinase ligand 5; Ephrin-B2; HTK ligand; HTK-L; LERK-5; ligand of eph-related kinase 5; MGC126226; MGC126227; MGC126228
Gene Aliases: EFNB2; ELF-2; Elf2; Epl5; EPLG5; Htk-L; HTKL; LERK-5; LERK5; NLERK-1
UniProt ID: (Human) P52799, (Mouse) P52800
Entrez Gene ID: (Human) 1948, (Mouse) 13642, (Rat) 306636
Molecular Function:
membrane-bound signaling molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support