Search Thermo Fisher Scientific
FIGURE: 1 / 5
The PA1-73003 antibody reacts with Epstein-Barr virus p18.
PA1-73003 has been successfully used in ELISA and immunofluorescence applications. IHC and WB are potential applications but have not yet been tested.
The PA1-73003 antibody was raised against Recombinant EBV p18.
Recommended short term storage (< 6 months) is at 4ºC. For longer term storage, aliquot and freeze.
Epstein-Barr virus (EBV) is a human herpes virus, which is associated with conditions such as Hodgkin's disease and Burkitt's Lymphoma and is the causative agent in mononucleosis in adolescents. EBV latently infects B lymphocytes. Infected B cells express EBV nuclear antigens and latent proteins LMP1, LMP2A and LMP2B. LMP2A forms aggregates in the plasma membranes of B lymphocytes, where it functions as a negative regulator of the Src and Syk protein tyrosine kinases. Studies show that LMP2A blocks B-cell receptor (BCR) signal transduction in EBV immortalized B cells in vitro and may play an important role in maintaining a latent EBV infection within the peripheral blood B cells of infected individuals.EBNA-1 is the only viral protein consistently expressed in all malignancies associated with EBV.
Protein Aliases: EBV p18