Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
QED Bioscience
Reconstitute in distilled water.
This gene encodes a member of a member of a high affinity glutamate transporter family. Mutations in this gene are associated with episodic ataxia, Type 6. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EAAT-1; Excitatory amino acid transporter 1; FLJ25094; GLAST; Glial glutamate transporter; glutamate transporter; glutamate/aspartate transporter; Sodium-dependent glutamate/aspartate transporter 1; Solute carrier family 1 member 3; solute carrier family 1, member 3
Gene Aliases: EAAT1; GLAST; GLAST-1; GluT-1; Slc1a3
UniProt ID: (Rat) P24942
Entrez Gene ID: (Rat) 29483
Molecular Function: primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support