Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: DLNAPGRATG AVSYTMLYNC LDFPAGVVPV TTVTAEDEAQ MEHYRGYFGD IWDKMLQKGM KKSVGLPVAV QCVALPWQEE LCLRFMREVE RLMTPEKQSS
Degrades bioactive fatty acid amides like oleamide, the endogenous levels of cannabinoid, anandamide and myristic amide to their corresponding acids, thereby serving to terminate the signaling functions of these molecules. Hydrolyzes polyunsaturated substrate anandamide preferentially as compared to monounsaturated substrates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anandamide amidohydrolase 1; FA2H; Fatty acid ester hydrolase; Fatty-acid amide hydrolase 1; MGC102823; MGC138146; Oleamide hydrolase; Oleamide hydrolase 1
Gene Aliases: FAAH; FAAH-1; FAAH1; PSAB
UniProt ID: (Human) O00519
Entrez Gene ID: (Human) 2166
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support