Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
May play roles in fatty acid transport and compartmentalization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FABPI; fatty acid binding protein 1; fatty acid binding protein 2, intestinal; Fatty acid-binding protein 2; Fatty acid-binding protein, intestinal; I-FABP; intestinal fatty acid binding protein; intestinal fatty acid binding protein 2; Intestinal-type fatty acid-binding protein; MGC133132
Gene Aliases: FABP; FABP2; FABPI; I-FABP
UniProt ID: (Human) P12104, (Pig) Q45KW7, (Mouse) P55050, (Rat) P02693
Entrez Gene ID: (Human) 2169, (Pig) 595106, (Rabbit) 100354372, (Mouse) 14079, (Rat) 25598
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support