Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) FABP2, 3, 4, 5, 6, 7 and 9 is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene localizes to myelin sheaths of the peripheral nervous system. The encoded protein can bind both the membrane layers of the sheaths and monomeric lipids, and is thought to provide stability to the sheath. A defect in this gene was shown to be a cause of dominant demyelinating CMT neuropathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: M FABP8; Myelin P2 protein; Peripheral myelin protein 2
Gene Aliases: FABP8; M-FABP; MP2; P2; PMP2
UniProt ID: (Human) P02689
Entrez Gene ID: (Human) 5375
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support