Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNGLSLSEL CCLFCCPPCP GRIAAKLAFL PPEATYSLVP EPEPGPGGAG AAPLGTLRAS SGAPGRWKLH LTERADFQYS QRELDTIEVF PTKSARGNHV SCMYVRCVPG ARYTVLFSHG NAVDLGQMSS FYIGLGSRLH CNIFSYDYSG YGASSGRPSE RNLYADIDAA WQALRTRYGI SPDSIILYGQ SIGTVPTVDL ASRYECAAVV LHSPLTSGMR VAFPDTKKTY CFDAFPK (1-236 aa encoded by BC000158)
Hydrolyzes fatty acids from S-acylated cysteine residues in proteins (PubMed:26701913). Has depalmitoylating activity towards NRAS (PubMed:26701913). Has depalmitoylating activity towards DLG4/PSD95 (PubMed:26701913). May have depalmitoylating activity towards MAP6. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Abhydrolase domain-containing protein 17A; abhydrolase domain-containing protein FAM108A; abhydrolase domain-containing protein FAM108A1; Alpha/beta hydrolase domain-containing protein 17A; family with sequence similarity 108, member A; family with sequence similarity 108, member A1; mFLJ00358 protein
Gene Aliases: 1700013O15Rik; ABHD17A; BC005632; C19orf27; D10Bwg1364e; Fam108a; FAM108A1; RGD1359682; upf0227
UniProt ID: (Human) Q96GS6, (Mouse) Q99JW1, (Rat) Q5XIJ5
Entrez Gene ID: (Human) 81926, (Mouse) 216169, (Rat) 299617
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support