Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QRKHLSSSS RAGHQEKEQY SRLSGGGHYN SQKTLSTSSI QNEIPKKKSK FESITTNGDS FSPDLALDSP GTDHFVIIAQ LKEEVAT (142-227 aa encoded by BC025768 )
FAM76A is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 76, member A; Protein FAM76A
Gene Aliases: FAM76A; RGD1305162
UniProt ID: (Human) Q8TAV0, (Mouse) Q922G2
Entrez Gene ID: (Human) 199870, (Rat) 362618, (Mouse) 230789
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support