Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AQQQTQLNL RSLLVNPEGP TLMRLNSVQS SERPLFLVHP IEGSTTVFHS LASRLSIPTY GLQCTRAAPL DSIHSLAAYY IDCIRQVQPE GPYRVAGYSY GACVAFEMCS QLQAQQSPAP THNSLFLFDG SPTYVLAYTQ SYRAKLTPGC EAEAETEAIC FFVQQFTDME HNRVLEALLP LKGLEERVAA AVDLIIKSHQ GLDRQELSFA ARSFYYKLRA AEQYTPKAKY HGNVMLLRAK TGGAYGEDLG ADYNLSQVCD GKVSVHVIEG DHRTLLEGSG LESIISIIHS SLAEPRVSVR EG (139-439 aa encoded by BC007909)
Fatty acid synthase is a multifunctional protein. Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA, in the presence of NADPH, into long-chain saturated fatty acids. In some cancer cell lines, this protein has been found to be fused with estrogen receptor-alpha (ER-alpha), in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-alpha.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fatty acid synthase; MGC14367; MGC15706; short chain dehydrogenase/reductase family 27X, member 1; Type I fatty acid synthase
Gene Aliases: FAS; FASN; OA-519; SDR27X1
UniProt ID: (Human) P49327
Entrez Gene ID: (Human) 2194
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support