Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 12
Positive Control: 293T, A431, HepG2, Neuro 2A, GL261, PC-12, Rat2
Predicted Reactivity: Pig (80%), Bovine (81%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Fatty acid synthase is a multifunctional protein. Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA, in the presence of NADPH, into long-chain saturated fatty acids. In some cancer cell lines, this protein has been found to be fused with estrogen receptor-alpha (ER-alpha), in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-alpha.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fatty acid synthase; MGC14367; MGC15706; short chain dehydrogenase/reductase family 27X, member 1; Type I FAS; Type I Fatty Acid Synthase
Gene Aliases: A630082H08Rik; FAS; FASN; OA-519; SDR27X1
UniProt ID: (Human) P49327, (Mouse) P19096, (Rat) P12785
Entrez Gene ID: (Human) 2194, (Mouse) 14104, (Rat) 50671
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support