Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LGKKPKAPAL RFLKRTLSNE SEESVKSTTL AVDYPKTPTG SPATEVSAKW THLTEFELKG LKALVEKLES LPENKKCVPE GIEDPQALLE GVKNVLKEH
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs, which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [Histone-H3]-lysine-36 demethylase 1B; CXXC-type zinc finger protein 2; F-box and leucine-rich repeat protein 10; F-box protein FBL10; F-box/LRR-repeat protein 10; JEMMA (Jumonji domain, EMSY-interactor, methyltransferase motif) protein; JmjC domain-containing histone demethylation protein 1B; jumonji C domain-containing histone demethylase 1B; Jumonji domain-containing EMSY-interactor methyltransferase motif protein; lysine (K)-specific demethylase 2B; Lysine-specific demethylase 2B; Protein JEMMA; Protein-containing CXXC domain 2
Gene Aliases: CXXC2; FBL10; FBXL10; JHDM1B; KDM2B; NDY1; PCCX2
UniProt ID: (Human) Q8NHM5
Entrez Gene ID: (Human) 84678
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support