Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. This protein is highly similar to the rat NFB42 (neural F Box 42 kDa) protein which is enriched in the nervous system and may play a role in maintaining neurons in a postmitotic state.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: F-box gene 1; F-box only protein 2; neural F box protein NFB42; organ of Corti protein 1; prion protein ligand 4
Gene Aliases: FBG1; Fbs1; Fbs2; FBX2; FBXO2; NFB42; OCP1; Prpl4
UniProt ID: (Human) Q9UK22, (Mouse) Q80UW2
Entrez Gene ID: (Human) 26232, (Mouse) 230904, (Rat) 85273
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support