Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MDGDGDPESV GQPEEASPEE QPEEASAEEE RPEDQQEEEA AAAAAYLDEL PEPLLLRVLA ALPAAELVQA CRLVCLRWKE LVDGAPLWLL KCQQEGLVPE GGVEEERDHW QQFYFLSKRR RNLLRNPCGE EDLEGWCDVE HGGDGWRVEE LPGDSGVEFT HDESVKKYFA SSFEWCRKAQ VIDLQAEGYW EELLDTTQPA IVVKDWYSGR SDAGCLYELT VKLLSEHENV LAEFSSGQVA VPQDSDGGGW MEISHTFTDY GPGVRFVRFE HGGQDSVYWK GWFGARVTNS SVWVEP
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. This protein is highly similar to the rat NFB42 (neural F Box 42 kDa) protein which is enriched in the nervous system and may play a role in maintaining neurons in a postmitotic state.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: F-box gene 1; F-box only protein 2; organ of Corti protein 1
Gene Aliases: FBG1; Fbs1; FBX2; FBXO2; NFB42; OCP1
UniProt ID: (Human) Q9UK22
Entrez Gene ID: (Human) 26232
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support