Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Members of the F-box protein family, such as FBXO42, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: F-box only protein 42; Just one F-box and Kelch domain-containing protein
Gene Aliases: 6720460I06Rik; FBX42; FBXO42; JFK; KIAA1332; mKIAA1332
UniProt ID: (Human) Q6P3S6, (Mouse) Q6PDJ6
Entrez Gene ID: (Human) 54455, (Rat) 362646, (Mouse) 213499
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support