Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASSSDSEDD SFMAVDQEET VLEGTMDQDE EPHPVLEAEE TRHNRSMSEL PEEVLEYILS FLSPYQEHKT AALVCKQWYR LIKGVAHQCY HGFMKAVQEG NIQWESRTYP YPGTPITQRF SHSACYYDAN QSMYVFGGCT QSSCNAAFND LWRLDLNSKE WIRPLASGSY PSPKAGATLV VYKDLLVLFG GWTRPSPYPL HQPERFFDEI HTYSPSKNWW NCIVTTHGPP PMAGHSSCVI DDKMIVFGGS LGSRQMSNDV WVLDLEQWAW SKPNISGPSP HPRGGQSQIV IDDATILILG GCGGPNALFK DAWLLHMHSG PWAWQPLKVE NEEHGAPELW CHPACRVGQC VVVFSQAPSG RAPLSPSLNS RPSPISATPP ALVPETREYR SQSPVRSMDE APCVNGRWGT LRPRAQRQTP SGSREGSLSP ARGDGSPILN GGSLSPGTAA VGGSLDSPVQ AISPSTPSAP EGYDLKIGLS LAPRRGSLPD QKDLRLGSID LNWDLKPASS SNPMDGMDNR TVGGSMRHPP EQTNGVHTPP HVASALAGAV SPGALRRSLE AIKAMSSKGP SASAALSPPL GSSPGSPGSQ SLSSGETVPI PRPGPAQGDG HSLPPIARRL GHHPPQSLNV GKPLYQSMNC KPMQMYVLDI KDTKEKGRVK WKVFNSSSVV GPPETSLHTV VQGRGELIIF GGLMDKKQNV KYYPKTNALY FVRAKR
Members of the F-box protein family, such as FBXO42, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: F-box only protein 42; Just one F-box and Kelch domain-containing protein
Gene Aliases: FBX42; FBXO42; JFK; KIAA1332
UniProt ID: (Human) Q6P3S6
Entrez Gene ID: (Human) 54455
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support