Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%
Component of E3 ubiquitin ligase complexes. Required for normal neuromuscular synaptogenesis, axon pathfinding and neuronal migration. Plays a role in the regulation of neurotransmission at mature neurons. May controls synaptic activity by controlling UNC13A via ubiquitin dependent pathway. Specifically recognizes TP73, promoting its ubiquitination and degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: F-box only protein 45; F-box/SPRY domain-containing protein 1; hFbxo45
Gene Aliases: FBX45; FBXO45
UniProt ID: (Human) P0C2W1
Entrez Gene ID: (Human) 200933
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support