Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSRRPCSCA LRPPRCSCSA SPSAVTAAGR PRPSDSCKEE SSTLSVKMKC DFNCNHVHSG LKLVKPDDIG RLVSYTPAYL EGSCKDCIKD YERLSCIGSP IVSPRIVELE TESKRLHNKE NQHVQQTLNS TNEIEALETS RLYEDSGYSS FSLQSGLSEH EEGSLLEENF GDSLQSCLLQ IQSPDQYPNK NLLPVLHFEK VVCSTLKKNA KRNPKVDREM LKEIIARGNF RLQNIIGRKM GLECVDILSE LFRRGLRHVL ATILAQLSDM DLINVSKVST TWKKILEDDK GAFQLYSKAI QRVTENNNKF SPHASTREYV MFRTPLASVQ KSAAQTSLKK DAQTKLSNQG DQKGSTYSRH NEFSEVAKTL KKNESLKACI RCNSPAKYDC YLQRATCKRE GCGFDYCTKC LCNYHTTKDC SDGKLLKASC KIGPLPGTKK SKKNLRRL (1-447 aa encoded by BC018905)
F-box proteins, critical components of the conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Novel human F-box proteins named FBG1, FBG2, FBG3, FBG4 and FBG5 constitute a subfamily of mammalian F-box proteins. FBG genes are expressed in a limited number of human tissues including kidney, liver, brain and muscle tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Early mitotic inhibitor 1; f-box only protein 31; F-box only protein 5; F-box protein Fbx5; fbxo5
Gene Aliases: 2510044I10Rik; C85305; EMI1; FBX5; Fbxo31; FBXO5
UniProt ID: (Human) Q9UKT4, (Mouse) Q7TSG3
Entrez Gene ID: (Human) 26271, (Mouse) 67141, (Rat) 292263
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support